Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human NT-4, Animal-Free Recombinant Protein, PeproTech®

Catalog No. p-7246100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF45004100 100 μg
AF4500410UG 10 μg
AF450041MG 1 mg
AF45004500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF45004100 Supplier Gibco™ Supplier No. AF45004100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human NT-4 is a noncovalently linked homodimer of two 14.0 kDa polypeptide monomers (260 total amino acid residues). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P34130
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 4909
Molecular Weight (g/mol) 28.1 kDa
Name Human NT-4, Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias 2900040K06Rik; AI462899; GLC10; GLC1O; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 4; Neurotrophin 4/5; neurotrophin 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); neurotrophin-4; Neurotrophin-5; neutrophic factor 4; Neutrophic factor 5; NT4; NT-4; NT4/5; NT-4/5; NT4/NT5; NT4P; NT5; NT-5; Ntf4; Ntf5; Ntf-5
Common Name NTF4
Gene Symbol NTF4
Biological Activity Determined by its ability to bind recombinant human TrkB Fc Chimera in a functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Sequence GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.