Learn More
Sino Biological Anti-5'-nucleotidase/CD73/NT5E Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10904MM07F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD73 / NT5E (rh CD73 / NT5E; Catalog#10904-H08H; NP_002517.1; Met1-Lys547) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8145