Learn More
Sino Biological Recombinant Human 5'-nucleotidase/CD73/NT5E Protein (hFc Tag)
Supplier:  Sino Biological 10904H02H100
Encompass_Preferred
A DNA sequence encoding the human NT5E (NP_002517.1, with natural variant Thr 376 Ala) (Met1-Lys547) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5079