Learn More
Sino Biological Recombinant Human 5'-nucleotidase/CD73/NT5E Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10904H08H1
Encompass_Preferred
A DNA sequence encoding the human NT5E (NP_002517.1) (Met1-Lys547) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5078