Learn More
Sino Biological Anti-TrkA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11073MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TrkA / NTRK1 (rh TrkA / NTRK1; Catalog#11073-H08H; NP_002520.2; Met1-Pro382). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0365