Learn More
Sino Biological Anti-TrkA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11073RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NTRK1 / Trk-A (rh NTRK1; Catalog#11073-H08H; NP_002520.2; Met 1-Pro 382). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8719