Learn More
Sino Biological Recombinant Human TrkA Protein (aa 285-413, His Tag)
Supplier:  Sino Biological 11073H07E11
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Pro 285-Glu 413) of human NTRK1 (NP_002520.2), corresponding to the Ig-like C2-type 2 domain, was expressed and purified, with a N-terminal polyhistidine tag.
Catalog No. 50-255-4883