Learn More
Sino Biological Recombinant Human TrkA Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11073H41HB20
Encompass_Preferred
A DNA sequence encoding the Human NTRK1 (NP_002520.2) (Met1-Gly423) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4881