Learn More
Sino Biological Anti-TrkB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10047T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TrkB (rh TrkB; Catalog#10047-H08H; NP_001007098.1; Met1-His430). TrkB specific IgG was purified by Human TrkB affinity chromatography.
Catalog No. 50-241-1055