Learn More
Sino Biological Human NTRK2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10047RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NTRK2 / Trk-B extracellular domain (rh Trk-B; Catalog#10047-H08H; NP_001007098.1; Met 1-His 430). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1745