Learn More
Sino Biological Recombinant Human TrkB Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10047H08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-His430) of human TrkB / NTRK2 (NP_001007098.1) precursor was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5515