Learn More
Sino Biological Anti-TrkC Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10048MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Trk-C / NTRK3 (rh Trk-C; Catalog#10048-H08H; NP_001007157.1; Met1-Asp428). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8917