Learn More
Biomatik Corporation Human Nuclear Distribution Gene C Homolog (NUDC) ELISA Kit
Supplier:  Biomatik Corporation EKU06347
Encompass_Preferred
Human Nuclear Distribution Gene C Homolog (NUDC) ELISA Kit for detecting NUDC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y266. Gene ID: 10726. Target Synonyms: HNUDC; MNUDC; NPD011. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5888