Learn More
Biomatik Corporation Human Nuclear Factor Kappa B (NFkB) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11974
Encompass_Preferred
Human Nuclear Factor Kappa B (NFkB) Mini sample ELISA Kit for detecting NFkB in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19838. Gene ID: 4790. Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5272