Learn More
Biomatik Corporation Human Nuclear Factor Kappa B2 (NFkB2) ELISA Kit
Supplier:  Biomatik Corporation EKU06355
Encompass_Preferred
Human Nuclear Factor Kappa B2 (NFkB2) ELISA Kit for detecting NFkB2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00653. Gene ID: 4791. Target Synonyms: LYT-10; H2TF1; Lymphocyte translocation chromosome 10; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 2 (p49/P100) ; Nuclear factor NF-kappa-B p52. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6738