Learn More
Biomatik Corporation Human Nuclear Receptor Corepressor 1 (NCOR1) ELISA Kit
Supplier:  Biomatik Corporation EKU11321
Encompass_Preferred
Human Nuclear Receptor Corepressor 1 (NCOR1) ELISA Kit for detecting NCOR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75376. Gene ID: 9611. Target Synonyms: KIAA1047; N-CoR; TRAC1; hN-CoR; Thyroid Hormone And Retinoic Acid Receptor Associated Corepressor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6203