Learn More
Biomatik Corporation Human Nuclear Receptor Subfamily 0,Group B,Member 2 (NR0B2) ELISA Kit
Supplier:  Biomatik Corporation EKU07375
Encompass_Preferred
Human Nuclear Receptor Subfamily 0,Group B,Member 2 (NR0B2) ELISA Kit for detecting NR0B2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15466. Gene ID: 8431. Target Synonyms: SHP; SHP1; Small Heterodimer Partner; Orphan nuclear receptor SHP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6716