Learn More
Biomatik Corporation Human Nuclear Transcription Factor Y Gamma (NFYC) ELISA Kit
Supplier:  Biomatik Corporation EKU06370
Encompass_Preferred
Human Nuclear Transcription Factor Y Gamma (NFYC) ELISA Kit for detecting NFYC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13952. Gene ID: 4802. Target Synonyms: CBF-C; NFYg; CBFC; H1TF2A; HAP5; HSM; NF-YC; hCBF-C; CAAT box DNA-binding protein subunit C; Nuclear transcription factor Y subunit C; Transactivator HSM-1/2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5660