Learn More
Biomatik Corporation Human Nucleobindin 1 (NUCB1) ELISA Kit
Supplier:  Biomatik Corporation EKU11735
Encompass_Preferred
Human Nucleobindin 1 (NUCB1) ELISA Kit for detecting NUCB1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02818. Gene ID: 4924. Target Synonyms: NUC; CALNUC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7152