Learn More
Biomatik Corporation Human Nucleobindin 2 (NUCB2) ELISA Kit
Supplier:  Biomatik Corporation EKU12476
Encompass_Preferred
Human Nucleobindin 2 (NUCB2) ELISA Kit for detecting NUCB2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80303. Gene ID: 4925. Target Synonyms: NUCB2; NEFA; Nucleobindin 2; DNA-binding protein NEFA; Gastric cancer antigen Zg4; Prepronesfatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6025