Learn More
ACROBiosystems ACROBiosystems Human Nucleolin / NCL Protein, Fc Tag
Supplier:  ACROBiosystems NULH52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human Nucleolin, Fc Tag (NUL-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accession # P19338-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 102.9 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6926