Learn More
ACROBiosystems ACROBiosystems Human Nucleolin / NCL Protein, Mouse IgG2a Fc Tag
Catalog No. 502106925
Encompass_Preferred
Recombinant Protein;100UG;Human Nucleolin, Mouse IgG2a Fc Tag (NUL-H5254) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accession # P19338-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 103.5 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6925 Supplier ACROBiosystems Supplier No. NULH5254100UG
Recombinant Protein;100UG;Human Nucleolin, Mouse IgG2a Fc Tag (NUL-H5254) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Glu 710 (Accession # P19338-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 103.5 kDa. The protein migrates as 130-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.