Learn More
Biomatik Corporation Human Nucleophosmin 3 (NPM3) ELISA Kit
Supplier:  Biomatik Corporation EKU12335
Encompass_Preferred
Human Nucleophosmin 3 (NPM3) ELISA Kit for detecting NPM3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75607. Gene ID: 10360. Target Synonyms: PORMIN; TMEM123; Nucleophosmin/Nucleoplasmin 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5920