Learn More
Abcam Human Nucleosome protein
Catalog No. 502704057
Encompass_Preferred
Human Nucleosome protein is a Full Length protein, expressed in HEK 293, with >=90% purity and suitable for SDS-PAGE.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4057 Supplier Abcam Supplier No. AB271797600UG
Human Nucleosome protein is a Full Length protein, expressed in HEK 293, with >=90% purity and suitable for SDS-PAGE.
The product is subject to the following: Abcam Restricted Use Statement