Learn More
Biomatik Corporation Human Nucleotide Binding Oligomerization Domain Containing Protein 2 (NOD2) CLIA Kit
Supplier:  Biomatik Corporation EKU08960
Encompass_Preferred
Human Nucleotide Binding Oligomerization Domain Containing Protein 2 (NOD2) CLIA Kit for detecting NOD2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 17.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9HC29. Gene ID: 64127. Target Synonyms: CD; ACUG; BLAU; CARD15; CLR16.3; IBD1; NLRC2; NOD2B; PSORAS1; Caspase recruitment domain-containing protein 15; Inflammatory bowel disease protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7797