Learn More
Sino Biological Human NUDT2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10944MM06T100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NUDT2 (rh NUDT2; Catalog#12551-H07E; P50583; Met1-Ala147). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4238