Learn More
Sino Biological Anti-Nup107 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102135T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nup107, and purified by antigen affinity chromatography.
Catalog No. 50-240-9772