Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01 NY-ESO-1 (SLLMWITQV) Complex Protein (HL1-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) & SLLMWITQV & Ile 21 - Met 119 (B2M) (Accession # AAA59606.1 (HLA-A*020
Catalog No. 502253854
Encompass_Preferred
NY-ESO-1, which is also well-known as New York esophageal squamous cell carcinoma 1, is an efficient target for cancer immunotherapy. This antigen is a member of cancer-testis antigens (CTAs) and is highly expressed in various cancers, including melanoma, ovarian, cervical cancer, etc. Adoptive T cell therapy with HLA-A2 restricted NY-ESO-1 transduced CD8+ T cells has improved the clinical response rates and overall survival of treatment-refractory melanoma patients.
This item is not returnable. View return policy
Catalog No. 50-225-3854 Supplier ACROBiosystems Supplier No. HL1H82E425UG
NY-ESO-1, which is also well-known as New York esophageal squamous cell carcinoma 1, is an efficient target for cancer immunotherapy. This antigen is a member of cancer-testis antigens (CTAs) and is highly expressed in various cancers, including melanoma, ovarian, cervical cancer, etc. Adoptive T cell therapy with HLA-A2 restricted NY-ESO-1 transduced CD8+ T cells has improved the clinical response rates and overall survival of treatment-refractory melanoma patients.