Learn More
Raybiotech Inc C-Series Human Obesity Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHADI18
Encompass_Preferred
Detects 62 Human Adipokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-ADI-1-8
Catalog No. 50-194-8455