Learn More
Raybiotech Inc G-Series Human Obesity Array 3 Kit.
Supplier:  Raybiotech Inc GSHADI32
Encompass_Preferred
Semi-quantitative measurement of 40 human adipokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Adiponectin (ACRP30), Adipsin (Complement Factor D), AgRP, ANGPTL4, BDNF, Chemerin, CRP (C-Reactive Protein), Growth Hormone, IFN-gamma, IGFBP-1, IGFBP-2, IGF-1, IL-10, IL-12 p40, IL-12 p70, IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-6, IL-8 (CXCL8), Insulin, Leptin, Lipocalin-2 (NGAL), MSP alpha/beta, Osteoprotegerin (TNFRSF11B), PAI-1, PDGF-BB, Pepsinogen 1, Pepsinogen 2, Procalcitonin, Prolactin, RANTES (CCL5), RBP4, Resistin, SAA (Serum Amyloid A), TGF beta 1, Thrombospondin 1, TNF RI (TNFRSF1A), TNF RII (TNFRSF1B), TNF alpha, VEGF-A
Catalog No. 50-195-0355