Learn More
Raybiotech Inc Quantibody Human Obesity Array 1 Kit.
Supplier:  Raybiotech Inc QAHADI11
Encompass_Preferred
Detects 10 Human Adipokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IGF-1, IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), Insulin, Leptin, MCP-1 (CCL2), PAI-1, Resistin, TNF alpha
Catalog No. 50-195-0785