Learn More
Raybiotech Inc Human OCT-4 ELISA Kit
Supplier:  Raybiotech Inc ELHOCT41
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.20 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ALK-3 (BMPR-IA), ALK-6 (BMPR-IB), Brachyury, CD38, E-Cadherein, GATA4, hCGbeta, Nanog, Nestin, PDX-1 (PF-1), SOX17, SOX2, VEGFR2
Catalog No. 50-194-9602