Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human OCT2 (G) SLC Transporter Cells

Catalog No. GM1105G
Encompass_Preferred
Change view
Click to view available options
Quantity:
8–10M cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GM1105G 8–10M cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GM1105G Supplier Gibco™ Supplier No. GM1105G
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

HEK293 cells that transiently overexpress the OCT2 solute carrier (SLC) transport protein

  • TRANSiPORT Human OCT2 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OCT2 solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The OCT2 SLC transporter protein is found in kidney proximal tubule cells.
  • Assess potential for transporter-mediated drug metabolism
  • Easy to use format
  • Obtain high quality results with a large signal-to-noise ratio
  • Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
  • Mock HEK293 cells (Cat. No. GM1101G) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
  • The convenient product format enables data generation in 2 days from thawing of the cells to final results
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen vapor phase.
Cell Line 293 (HEK)
Culture Type Mammalian Cell Culture
Species Human
Product Type SLC Transporter Cell
Quantity 8–10M cells
What are TRANSiPORT human solute carrier (SLC) transporter cells?

TRANSiPORT human SLC transporter cells are HEK293 cells that transiently overexpress a solute carrier (SLC) transporter protein. Solute carriers are one of two superfamilies of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes into and out of cells. Membrane transporters are found in hepatic cells, kidney proximal tubule cells, intestinal cells, and brain cells.

Is there a protocol available for the TRANSiPORT solute carrier (SLC) transporter cells?

Yes, the protocol is available here (https://www.genomembrane.com/information).

What should I use as a control with TRANSiPORT solute carrier (SLC) transporter cells?

Mock HEK293 cells (Cat. No. GM1001) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of SLC transport due to the presence of endogenous transporters or non-specific binding.

With TRANSiPORT solute carrier (SLC) transporter cells, what should my choice of measurement be for SLC transporter activity?

Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.

How many cells can I expect in one vial of TRANSiPORT solute carrier (SLC) transporter cells?

One vial contains 8-12 million cells with high viability, designed to seed one 24-well plate for single use.

What are the shipping and storage conditions for TRANSiPORT solute carrier (SLC) transporter cells?

Domestic shipments are on dry ice and shipped overnight. We ask our customers to store the product under liquid nitrogen after they receive it. International shipments are under liquid nitrogen conditions.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.