Learn More
IPHASE PHARMA SERVICES LLC Human OCT2 SLC Transporter Cells,8-10million
Catalog No. NC3966937
Encompass_Preferred
Participate in drug-drug interactions and drug uptake/ drug efflux. SLC transporter developed using transfected cells as a method and HEK293F cells as a vector.They play key roles in drug resistance, metabolism, and cellular detoxification and usually be used in DDI studies. The product need to be used in conjunction with MOCK/HEK293F Cell, Transporter Buffer and Polylysine Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3966937 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 01942A1.01
Participate in drug-drug interactions and drug uptake/ drug efflux. SLC transporter developed using transfected cells as a method and HEK293F cells as a vector.They play key roles in drug resistance, metabolism, and cellular detoxification and usually be used in DDI studies. The product need to be used in conjunction with MOCK/HEK293F Cell, Transporter Buffer and Polylysine Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.