Learn More
Sino Biological Recombinant Human Olfactomedin-4/OLFM4 Protein (His Tag)
Supplier:  Sino Biological 11639H08H50
Encompass_Preferred
A DNA sequence encoding the human OLFM4 (NP_006409.3) (Met 1-Gln 510) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6979