Learn More
Sino Biological Recombinant Anti-LOX-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10585R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human OLR1 / LOX1 (rh OLR1 / LOX1; Catalog#10585-H07H; P78380; Ser61-Gln273).
Catalog No. 50-240-9040