Learn More
Sino Biological Anti-LOX-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10585T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human OLR1 / LOX1 (rh OLR1 / LOX1; Catalog#10585-H07H; P78380; Ser61-Gln273). OLR1 / LOX1 specific IgG was purified by Human OLR1 / LOX1 affinity chromatography.
Catalog No. 50-240-9282