Learn More
Sino Biological Anti-LOX-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10585RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human OLR1 / LOX1 (rh OLR1 / LOX1; Catalog#10585-H07H; P78380; Ser 61-Gln 273). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0403