Learn More
Biocare Medical, LLC Human-on-Human HRP-Polymer Kit, Enables Rapid Screening of Multiple Human Primary Antibody Clones
SDP
Catalog No. NC1071061
Encompass_Preferred
The Human-on-Human HRP-Polymer kit provides reagents to tag the human primary antibody of interest with digoxigenin and detect it on human tissue with an HRPpolymer system. With this detection system, human antibodies bound to human tissue can be detected with high specificity and sensitivity, while significantly reducing or eliminating background.
Catalog No. NC1071061 Supplier Biocare Medical, LLC Supplier No. BRR4056KG
The Human-on-Human HRP-Polymer kit provides reagents to tag the human primary antibody of interest with digoxigenin and detect it on human tissue with an HRPpolymer system. With this detection system, human antibodies bound to human tissue can be detected with high specificity and sensitivity, while significantly reducing or eliminating background.