Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Oncostatin M (196 aa) Recombinant Protein, PeproTech®

Catalog No. 30010H10UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30010H10UG 10 μg
30010H100UG 100 μg
300-10H-1MG 1 mg
30010H250UG 250 μg
300-10H-2UG 2 μg
30010H500UG 500 μg
30010H50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 30010H10UG Supplier Gibco™ Supplier No. 30010H10UG
  • $225.00
    $168.65 / Each of 1

    Save $56.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

PeproTech's Recombinant Human Oncostatin M (196 a.a.) is a HEK293 cell-derived, 196 amino-acid length glycoprotein that has a calculated theoretical molecular weight of 22.2 kDa, but migrates at an apparent molecular weight of 32-34 kDa by SDS-PAGE analysis under reducing conditions due to glycosylation. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi's sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi's sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13725
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 5008
Molecular Weight (g/mol) 22.2 kDa
Name Human Oncostatin M (196 aa)
Purification Method Purified
Quantity 10 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias MGC20461; OncoM; oncostatin M; oncostatin-M; OSM
Common Name Oncostatin M
Gene Symbol OSM
Biological Activity Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSRR
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of human TF-1 cells. The expected ED50 is ≤ 2.0 ng/ml, corresponding to a specific activity of ≥ 5 x 10^5 units/mg.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.