Learn More
ACROBiosystems Human Oncostatin M (OSM) (26-221) Protein, Tag Free (MALS verified)
Catalog No. 502331679
Encompass_Preferred
Human Oncostatin M (OSM) (26-221), Tag Free (OSM-H5214) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Arg 221 (Accession # P13725).
Catalog No. 50-233-1679 Supplier ACROBiosystems Supplier No. OSMH521420UG
Human Oncostatin M (OSM) (26-221), Tag Free (OSM-H5214) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Arg 221 (Accession # P13725).