Learn More
Biomatik Corporation Human Oncostatin M (OSM) CLIA Kit
Supplier:  Biomatik Corporation EKU08831
Encompass_Preferred
Human Oncostatin M (OSM) CLIA Kit for detecting OSM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.93pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13725. Gene ID: 5008. Target Synonyms: OS-M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8618