Learn More
Sino Biological Anti-ONECUT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104590T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ONECUT1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8431