Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human OPA1 (aa 707-803) Control Fragment Recombinant Protein

Catalog No. RP96480
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96480 Supplier Invitrogen™ Supplier No. RP96480
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57874. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

OPA1 is a dynamin-related GTPase that is critical for the maintenance of mitochondrial morphology and mtDNA. The most commonly associated phenotype with OPA1 mutations is heterozygous optic atrophy, a heterozygous dominant trait that causes reduced visual clarity and sometimes blindness. The disease usually begins in childhood and increases in severity throughout the life of affected individuals. Usually, this phenotype is attributed to the degeneration of optic nerve fibers. Interestingly, the same type of nerve degeneration seems to be partially causative of certain schizophrenia characteristics. OPA1 dysfunction also seems to be implicated in this case; mitochondrial networks associated with critical nerves seem to link schizophrenia and OPA1. The dysfunction is associated with issues with apoptosis and normal cellular metabolic regulation, all regulated through OPA1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60313
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 4976
Name Human OPA1 (aa 707-803) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1200011N24Rik; AI225888; AI847218; BERHS; Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; EMBL:AAI11072.1}; hypothetical protein; KIAA0567; Large GTP-binding protein; largeG; lilr3; MGM1; mitochondrial dynamin-like GTPase; mitochondrial OPA1; mKIAA0567; MTDPS14; NPG; NTG; OPA1; opa1 {ECO:0000312; OPA1 mitochondrial dynamin like GTPase; OPA1, mitochondrial dynamin like GTPase; optic atrophy 1; optic atrophy 1 (autosomal dominant); optic atrophy 1 homolog; optic atrophy 1-like protein; optic atrophy protein 1; Optic atrophy protein 1 homolog; RCJMB04_1m16; RN protein
Common Name OPA1
Gene Symbol OPA1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ESIKRHKWNDFAEDSLRVIQHNALEDRSISDKQQWDAAIYFMEEALQARLKDTENAIENMVGPDWKKRWLYWKNRTQEQCVHNETKNELEKMLKCNE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.