Learn More
Biomatik Corporation Human Optineurin (OPTN) ELISA Kit
Supplier:  Biomatik Corporation EKU09133
Encompass_Preferred
Human Optineurin (OPTN) ELISA Kit for detecting OPTN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96CV9. Gene ID: 10133. Target Synonyms: NRP; FIP2; GLC1E; HIP7; HYPL; TFIIIA-INTP; Glaucoma 1,Open Angle,E (Adult-Onset) ; Huntingtin-interacting protein 7; Optic neuropathy-inducing protein; E3-14.7K-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7791