Learn More
Sino Biological Anti-ORM2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13299T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ORM2 (rh ORM2; Catalog#13299-H08H; P19652; Met1-Ser201). ORM2 specific IgG was purified by Human ORM2 affinity chromatography.
Catalog No. 50-255-4260