Learn More
American Research Products Inc Human Ornithine carbamoyltrans
Catalog No. 5000328038
Encompass
Human Ornithine carbamoyltransferase mitochondrial (OTC) ELISA kit from CUSABIO Detection Range 1 25 mU/mL-80 mU/mL Sensitivity 0 312 mU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28038 Supplier American Research Products Inc Supplier No. CSBEL017270HU
Human Ornithine carbamoyltransferase mitochondrial (OTC) ELISA kit from CUSABIO Detection Range 1 25 mU/mL-80 mU/mL Sensitivity 0 312 mU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich