Learn More
Sino Biological Recombinant Human Oncostatin M/OSM Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10452H49HB20
Encompass_Preferred
A DNA sequence encoding the Human OSM (NP_065391.1) (Met1-Arg221) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5295