Learn More
ACROBiosystems Human OSMR, His Tag (OSR-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Met 740 (Accession # Q99650-1).
Supplier:  ACROBiosystems OSRH52H9100UG
Encompass_Preferred
Human OSMR, His Tag (OSR-H52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Met 740 (Accession # Q99650-1).
Catalog No. 50-225-3360