Learn More
Sino Biological Anti-OSMR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11226RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human OSMR / IL31RB (rh OSMR; Catalog#11226-H08H; NP_003990.1; Met 1-Met 740). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1212